accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7133-37-1

IUPAC Name: (methylsulfanyl)cyclohexane
Synonyms: (methylsulfanyl)cyclohexane
Molecular Weight (g/mol): 130.25
Molecular Formula: C7H14S
InChi Key: QQBIOCGHCKNYGP-UHFFFAOYSA-N
SMILES: CSC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)