accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7143-82-0

IUPAC Name: 6-methyl-2,3-dihydropyridazin-3-one
Synonyms: 6-methyl-2H-pyridazin-3-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: QZWIXLPWMGHDDD-UHFFFAOYSA-N
SMILES: CC1=NNC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results