accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7144-05-0

IUPAC Name: 1-(piperidin-4-yl)methanamine
Synonyms: 4-piperidinemethanamine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: LTEKQAPRXFBRNN-UHFFFAOYSA-N
SMILES: NCC1CCNCC1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)