accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7147-89-9

IUPAC Name: 4-chloro-5-methyl-2-nitrophenol
Synonyms: 4-chloro-5-methyl-2-nitrophenol
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: JBMGJOKJUYGIJH-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(C=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results