CAS RN 7148-03-0
IUPAC Name:
4'-methyl-[1,1'-biphenyl]-2-carboxylic acid
Synonyms:
4'-methyl-[1,1'-biphenyl]-2-carboxylic acid
Molecular Weight (g/mol):
212.25
Molecular Formula:
C14H12O2
InChi Key:
ZSTUEICKYWFYIC-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)C1=CC=CC=C1C(O)=O
1
–
1
of
1
results
1
2-(p-Tolyl)benzoic Acid 98.0+%, TCI America™
CAS: 7148-03-0 Molecular Formula: C14H12O2 Molecular Weight (g/mol): 212.248 MDL Number: MFCD00045826 InChI Key: ZSTUEICKYWFYIC-UHFFFAOYSA-N PubChem CID: 81568 IUPAC Name: 2-(4-methylphenyl)benzoic acid SMILES: CC1=CC=C(C=C1)C2=CC=CC=C2C(=O)O