accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7148-07-4

IUPAC Name: 1-(cyclopent-1-en-1-yl)pyrrolidine
Synonyms: 1-(cyclopent-1-en-1-yl)pyrrolidine
Molecular Weight (g/mol): 137.23
Molecular Formula: C9H15N
InChi Key: KOFSFYBXUYHNJL-UHFFFAOYSA-N
SMILES: C1CCN(C1)C1=CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results