accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7152-24-1

IUPAC Name: 2-(methylsulfanyl)-1H-1,3-benzodiazole
Synonyms: 2-(methylsulfanyl)-1H-1,3-benzodiazole
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: OCKJFOHZLXIAAT-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC=CC=C2N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)