accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7154-34-9

IUPAC Name: 6-nitroquinazoline-2,4-diamine
Synonyms: 6-nitroquinazoline-2,4-diamine
Molecular Weight (g/mol): 205.18
Molecular Formula: C8H7N5O2
InChi Key: YZMJNZRTRWPJFY-UHFFFAOYSA-N
SMILES: NC1=NC(N)=C2C=C(C=CC2=N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results