accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7154-73-6

IUPAC Name: 2-(pyrrolidin-1-yl)ethan-1-amine
Synonyms: 1-pyrrolidineethanamine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: WRXNJTBODVGDRY-UHFFFAOYSA-N
SMILES: NCCN1CCCC1
Boiling Point 
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)