accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 716-39-2

IUPAC Name: 1H,3H-naphtho[2,3-c]furan-1,3-dione
Synonyms: naphtho[2,3-c]furan-1,3-dione
Molecular Weight (g/mol): 198.18
Molecular Formula: C12H6O3
InChi Key: IZJDCINIYIMFGX-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1C=C1C=CC=CC1=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results