accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N(R)HO(R)OHNOO

CAS RN 716-61-0

IUPAC Name: (1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
Synonyms: (1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
Molecular Weight (g/mol): 213.21
Molecular Formula: C9H13N2O4
InChi Key: OCYJXSUPZMNXEN-RKDXNWHRSA-O
SMILES: [NH3+][C@H](CO)[C@H](O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)