accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 716-79-0

IUPAC Name: 2-phenyl-1H-1,3-benzodiazole
Synonyms: 2-phenylbenzimidazole
Molecular Weight (g/mol): 194.24
Molecular Formula: C13H10N2
InChi Key: DWYHDSLIWMUSOO-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2N=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)