accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71605-72-6

IUPAC Name: 2,1,3-benzothiadiazole-5-carbaldehyde
Synonyms: 2,1,3-benzothiadiazole-5-carbaldehyde
Molecular Weight (g/mol): 164.18
Molecular Formula: C7H4N2OS
InChi Key: GEFIFDVQYCPLHC-UHFFFAOYSA-N
SMILES: O=CC1=CC2=NSN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results