accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH(S)(S)OHO

CAS RN 71629-07-7

IUPAC Name: 7-[(1S,2S)-2-(heptylamino)cyclopentyl]heptanoic acid
Synonyms: 7-[(1S,2S)-2-(heptylamino)cyclopentyl]heptanoic acid
Molecular Weight (g/mol): 311.51
Molecular Formula: C19H37NO2
InChi Key: RZZNGDIYQSCFGK-ROUUACIJSA-N
SMILES: CCCCCCCN[C@H]1CCC[C@@H]1CCCCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results