accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7163-25-9

IUPAC Name: ethyl 3-hydroxynaphthalene-2-carboxylate
Synonyms: ethyl 3-hydroxynaphthalene-2-carboxylate
Molecular Weight (g/mol): 216.24
Molecular Formula: C13H12O3
InChi Key: CVEOWBRXZJEZRQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(O)C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results