accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71643-66-8

IUPAC Name: 4-chloro-2-iodophenol
Synonyms: 4-chloro-2-iodophenol
Molecular Weight (g/mol): 254.45
Molecular Formula: C6H4ClIO
InChi Key: JKPLMQJLGBBFLO-UHFFFAOYSA-N
SMILES: OC1=C(I)C=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results