accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71658-33-8

IUPAC Name: 2-(4-methylphenyl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 2-(4-methylphenyl)-3,5-diphenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 348.83
Molecular Formula: C20H17ClN4
InChi Key: NCJWJQNOYLTOCG-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results