CAS RN 71666-95-0
CAS RN 71666-95-0
IUPAC Name:
2-amino-3-(1H-imidazol-5-yl)propanamide dihydrochloride
Synonyms:
2-amino-3-(3H-imidazol-4-yl)propanamide dihydrochloride
Molecular Weight (g/mol):
227.09
Molecular Formula:
C6H12Cl2N4O
InChi Key:
CRBYFJCXMZNLTO-UHFFFAOYNA-N
SMILES:
Cl.Cl.NC(CC1=CN=CN1)C(N)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals L-Histidinamide dihydrochloride, 95%, Thermo Scientific™
CAS: 71666-95-0 Molecular Formula: C6H12Cl2N4O Molecular Weight (g/mol): 227.089 MDL Number: MFCD00058559 InChI Key: CRBYFJCXMZNLTO-XRIGFGBMSA-N Synonym: h-his-nh2.2hcl,l-histidinamide dihydrochloride,h-his-nh2 2hcl,h-his-nh 2 hcl,c6h10n4o.2hcl,h-his-nh2 . 2 hcl,l-histidine amide dihydrochloride,l-histidinamide-hydrogen chloride 1/2,h-his-nh2 inverted exclamation mark currency 2 hcl,2s-2-amino-3-1h-imidazol-4-yl propanamide dihydrochloride PubChem CID: 56777274 IUPAC Name: (2S)-2-amino-3-(1H-imidazol-5-yl)propanamide;dihydrochloride SMILES: C1=C(NC=N1)CC(C(=O)N)N.Cl.Cl