accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7169-34-8

IUPAC Name: 2,3-dihydro-1-benzofuran-3-one
Synonyms: 3(2H)-benzofuranone
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: MGKPCLNUSDGXGT-UHFFFAOYSA-N
SMILES: O=C1COC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results