accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOO

CAS RN 7169-37-1

IUPAC Name: 7-methoxy-2,3-dihydro-1-benzofuran-3-one
Synonyms: 7-methoxy-2H-1-benzofuran-3-one
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: QSJBSTJSAZCHSA-UHFFFAOYSA-N
SMILES: COC1=CC=CC2=C1OCC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results