accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHH3COOOHH2OH2OCa

CAS RN 71720-56-4

IUPAC Name: bis(2-(3-phenoxyphenyl)propanoic acid) calcium dihydrate
Synonyms: bis(fenoprofen) calcium dihydrate
Molecular Weight (g/mol): 560.66
Molecular Formula: C30H32CaO8
InChi Key: LXVOQQSZQOHNFB-UHFFFAOYNA-N
SMILES: O.O.[Ca].CC(C(O)=O)C1=CC(OC2=CC=CC=C2)=CC=C1.CC(C(O)=O)C1=CC(OC2=CC=CC=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results