accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N

CAS RN 7175-49-7

IUPAC Name: N-cyclohexyl-N-ethylcyclohexanamine
Synonyms: N-cyclohexyl-N-ethylcyclohexanamine
Molecular Weight (g/mol): 209.38
Molecular Formula: C14H27N
InChi Key: XRKQMIFKHDXFNQ-UHFFFAOYSA-N
SMILES: CCN(C1CCCCC1)C1CCCCC1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results