accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CH3CONNNNNH2HOOPSOHO

CAS RN 71774-13-5

IUPAC Name: 6-(6-amino-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinine-2-thione; triethylamine
Synonyms: 6-(6-aminopurin-9-yl)-2,7-dihydroxy-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinine-2-thione; triethylamine
Molecular Weight (g/mol): 446.46
Molecular Formula: C16H27N6O5PS
InChi Key: OXIPZMKSNMRTIV-UHFFFAOYNA-N
SMILES: CCN(CC)CC.NC1=C2N=CN(C3OC4COP(O)(=S)OC4C3O)C2=NC=N1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results