accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFFFF

CAS RN 718-64-9

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol
Synonyms: 1,1,1,3,3,3-hexafluoro-2-phenylpropan-2-ol
Molecular Weight (g/mol): 244.14
Molecular Formula: C9H6F6O
InChi Key: IZPIZCAYJQCTNG-UHFFFAOYSA-N
SMILES: OC(C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)