CAS RN 71808-65-6
IUPAC Name:
methoxydimethyloctadecylsilane
Synonyms:
methoxydimethyloctadecylsilane
Molecular Weight (g/mol):
342.68
Molecular Formula:
C21H46OSi
InChi Key:
RUFRLNPHRPYBLF-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCC[Si](C)(C)OC
1
–
1
of
1
results
1
Methoxy(dimethyl)octadecylsilane 90.0+%, TCI America™
CAS: 71808-65-6 Molecular Formula: C21H46OSi Molecular Weight (g/mol): 342.68 MDL Number: MFCD00042920 InChI Key: RUFRLNPHRPYBLF-UHFFFAOYSA-N PubChem CID: 172903 IUPAC Name: methoxydimethyloctadecylsilane SMILES: CCCCCCCCCCCCCCCCCC[Si](C)(C)OC