accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHONH2Cl

CAS RN 718596-54-4

IUPAC Name: 2-amino-3-(2,5-dichlorophenyl)propanoic acid
Synonyms: 2-amino-3-(2,5-dichlorophenyl)propanoic acid
Molecular Weight (g/mol): 234.08
Molecular Formula: C9H9Cl2NO2
InChi Key: SDKIMTATYJGPBJ-UHFFFAOYNA-N
SMILES: NC(CC1=C(Cl)C=CC(Cl)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)