accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 718635-09-7

IUPAC Name: sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazol-1-ide
Synonyms: protonix|protium|pantoprazole sodium|citrel
Molecular Weight (g/mol): 405.35
Molecular Formula: C16H14F2N3NaO4S
InChi Key: YNWDKZIIWCEDEE-UHFFFAOYNA-N
SMILES: [Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results