accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71868-10-5

IUPAC Name: 2-methyl-1-[4-(methylsulfanyl)phenyl]-2-(morpholin-4-yl)propan-1-one
Synonyms: methylthiophenyl morpholino isobutanone
Molecular Weight (g/mol): 279.4
Molecular Formula: C15H21NO2S
InChi Key: LWRBVKNFOYUCNP-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)C(=O)C(C)(C)N1CCOCC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)