accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNN

CAS RN 71878-80-3

IUPAC Name: 1-(methoxymethyl)-1H-1,2,3-benzotriazole
Synonyms: 1-(methoxymethyl)-1,2,3-benzotriazole
Molecular Weight (g/mol): 163.18
Molecular Formula: C8H9N3O
InChi Key: QEIXBXXKTUNWDK-UHFFFAOYSA-N
SMILES: COCN1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results