accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 71885-51-3

IUPAC Name: diethyl [(oxan-2-yloxy)methyl]phosphonate
Synonyms: diethyl (oxan-2-yloxy)methylphosphonate
Molecular Weight (g/mol): 252.25
Molecular Formula: C10H21O5P
InChi Key: RUPVGTAIRUGQIQ-UHFFFAOYNA-N
SMILES: CCOP(=O)(COC1CCCCO1)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results