accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiNH2OH3COH3COSiO

CAS RN 71888-97-6

IUPAC Name: (3-aminopropyl)triethoxysilane; silanedione
Synonyms: aminopropyl triethoxysilane; silica
Molecular Weight (g/mol): 281.46
Molecular Formula: C9H23NO5Si2
InChi Key: JIBSXWZIQTVOKJ-UHFFFAOYSA-N
SMILES: O=[Si]=O.CCO[Si](CCCN)(OCC)OCC
Material 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)