accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7189-69-7

IUPAC Name: 1-(1H-imidazole-1-sulfonyl)-1H-imidazole
Synonyms: 1-(imidazole-1-sulfonyl)imidazole
Molecular Weight (g/mol): 198.2
Molecular Formula: C6H6N4O2S
InChi Key: ZLKNPIVTWNMMMH-UHFFFAOYSA-N
SMILES: O=S(=O)(N1C=CN=C1)N1C=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)