accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 719-22-2

IUPAC Name: 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 220.31
Molecular Formula: C14H20O2
InChi Key: RDQSIADLBQFVMY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (4)
Boiling Point 
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)