accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 719-54-0

IUPAC Name: 10-methyl-9,10-dihydroacridin-9-one
Synonyms: 10-methylacridin-9-one
Molecular Weight (g/mol): 209.25
Molecular Formula: C14H11NO
InChi Key: XUVKSPPGPPFPQN-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2C(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results