accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 7194-85-6

IUPAC Name: octatriacontane
Synonyms: N-octatriacontane
Molecular Weight (g/mol): 535.04
Molecular Formula: C38H78
InChi Key: BVKCQBBZBGYNOP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results