accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOOHHCl

CAS RN 71985-79-0

IUPAC Name: 2-(pyrrolidin-2-yl)acetic acid hydrochloride
Synonyms: pyrrolidin-2-ylacetic acid hydrochloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: VQDACVOAOJQTPR-UHFFFAOYNA-N
SMILES: Cl.OC(=O)CC1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results