accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3O(S)HNOOOO

CAS RN 71989-33-8

IUPAC Name: (2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms: (2S)-3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 382.44
Molecular Formula: C22H24NO5
InChi Key: REITVGIIZHFVGU-IBGZPJMESA-M
SMILES: CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (7)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)