CAS RN 71989-38-3
N-Fmoc-O-tert-butyl-L-tyrosine, 98%
CAS: 71989-38-3 Molecular Formula: C28H29NO5 Molecular Weight (g/mol): 459.54 MDL Number: MFCD00037129 InChI Key: JAUKCFULLJFBFN-KSYWNVGFNA-N Synonym: fmoc-tyr tbu-oh,fmoc-o-tert-butyl-l-tyrosine,fmoc-tyr but-oh,fmoc-tyr t-bu-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid,fmoc-tyr but,n-fmoc-o-tert-butyl-l-tyrosine,nalpha-fmoc-o-tert-butyl-l-tyrosine,n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine,2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid PubChem CID: 10895791 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid SMILES: CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
FMOC-O-tert-Butyl-L-tyrosine, 98%
CAS: 71989-38-3 Molecular Formula: C28H29NO5 Molecular Weight (g/mol): 459.54 MDL Number: MFCD00037129 InChI Key: JAUKCFULLJFBFN-KSYWNVGFNA-N Synonym: fmoc-tyr tbu-oh,fmoc-o-tert-butyl-l-tyrosine,fmoc-tyr but-oh,fmoc-tyr t-bu-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid,fmoc-tyr but,n-fmoc-o-tert-butyl-l-tyrosine,nalpha-fmoc-o-tert-butyl-l-tyrosine,n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine,2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid PubChem CID: 10895791 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid SMILES: CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine 98.0+%, TCI America™
CAS: 71989-38-3 Molecular Formula: C28H29NO5 Molecular Weight (g/mol): 459.54 MDL Number: MFCD00037129 InChI Key: JAUKCFULLJFBFN-KSYWNVGFNA-N Synonym: fmoc-tyr tbu-oh,fmoc-o-tert-butyl-l-tyrosine,fmoc-tyr but-oh,fmoc-tyr t-bu-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid,fmoc-tyr but,n-fmoc-o-tert-butyl-l-tyrosine,nalpha-fmoc-o-tert-butyl-l-tyrosine,n-9-fluorenylmethoxy carbonyl-o-tert-butyl-l-tyrosine,2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid PubChem CID: 10895791 IUPAC Name: (2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid SMILES: CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
Fmoc-Tyr tBu -OH, Advanced ChemTech
CAS: 71989-38-3 Molecular Formula: C28H29NO5 Molecular Weight (g/mol): 459.54 MDL Number: MFCD00037129 InChI Key: JAUKCFULLJFBFN-KSYWNVGFNA-N IUPAC Name: (2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid SMILES: CC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
Sigma Aldrich Fmoc-Tyr(Tbu)-Oh
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.