accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)OH(S)NH2OHO

CAS RN 72-19-5

IUPAC Name: 2-amino-3-hydroxybutanoic acid
Synonyms: threonine(L) DL-allothreonine
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: AYFVYJQAPQTCCC-UHFFFAOYNA-N
SMILES: CC(O)C(N)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (35)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (10)
  • (1)
  • (3)
  • (5)
Quantity 
  • (7)
  • (1)
  • (1)
  • (8)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
Grade 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (11)
  • (2)
Physical Form 
  • (3)
  • (7)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (20)
  • (16)
  • (1)
  • (4)
  • (2)
  • (5)
  • (12)
  • (3)
  • (2)
  • (9)
  • (3)
  • (12)
  • (4)
  • (35)
  • (3)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (10)
  • (1)
  • (3)
  • (5)
  • (7)
  • (1)
  • (1)
  • (8)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (11)
  • (2)
  • (3)
  • (7)
  • (3)