accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72-20-8

IUPAC Name: 3,4,5,6,13,13-hexachloro-10-oxapentacyclo[6.3.1.1³,⁶.0²,⁷.0⁹,¹¹]tridec-4-ene
Synonyms: endrin dieldrin (iso)
Molecular Weight (g/mol): 380.9
Molecular Formula: C12H8Cl6O
InChi Key: DFBKLUNHFCTMDC-UHFFFAOYNA-N
SMILES: ClC1=C(Cl)C2(Cl)C3C4CC(C5OC45)C3C1(Cl)C2(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results