accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)(S)CH3(S)(R)OHCH(R)

CAS RN 72-33-3

IUPAC Name: (1R,3aS,3bR,9bS,11aS)-1-ethynyl-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonyms: mestranol EE(sub3)ME caswell no. 547A ovastol menophase mestranol preparation (product) epa pesticide chemical code 115401 delta-MVE 3-methoxy-17α-ethynylestradiol mestranolum
Molecular Weight (g/mol): 310.44
Molecular Formula: C21H26O2
InChi Key: IMSSROKUHAOUJS-MJCUULBUSA-N
SMILES: COC1=CC=C2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@@H]3CCC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results