accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNNNH2OPOHOOPOOHHOHONaNa

CAS RN 72003-83-9

IUPAC Name: [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid disodium
Synonyms: deoxyadenosine diphosphate disodium
Molecular Weight (g/mol): 457.18
Molecular Formula: C10H15N5Na2O9P2
InChi Key: PMCITFSCGIPAKY-UHFFFAOYNA-N
SMILES: [Na].[Na].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP(O)(O)=O)O3)C2=NC=N1
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)