CAS RN 7206-28-2
IUPAC Name:
dodec-5-ene
Synonyms:
dodec-5-ene
Molecular Weight (g/mol):
168.32
Molecular Formula:
C12H24
InChi Key:
ZOKYTRIEIDWYSG-UHFFFAOYSA-N
SMILES:
CCCCCCC=CCCCC
1
–
1
of
1
results
1
5-Dodecene 98.0+%, TCI America™
CAS: 7206-28-2 Molecular Formula: C12H24 Molecular Weight (g/mol): 168.324 MDL Number: MFCD02093465 InChI Key: ZOKYTRIEIDWYSG-LUAWRHEFSA-N PubChem CID: 5364658 IUPAC Name: (Z)-dodec-5-ene SMILES: CCCCCCC=CCCCC