accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7207-49-0

IUPAC Name: (Z)-N-(octan-2-ylidene)hydroxylamine
Synonyms: (Z)-N-(octan-2-ylidene)hydroxylamine
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: GZRPVYSKBVDCBV-HJWRWDBZSA-N
SMILES: CCCCCC\C(C)=N/O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results