accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSNOHOHCl

CAS RN 720720-96-7

IUPAC Name: 5-methyl-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride
Synonyms: 5-methyl-4H,6H,7H-[1,3]thiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride
Molecular Weight (g/mol): 234.7
Molecular Formula: C8H11ClN2O2S
InChi Key: ZWIYEBIMFPQYDI-UHFFFAOYSA-N
SMILES: Cl.CN1CCC2=C(C1)SC(=N2)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results