accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSBr

CAS RN 7209-11-2

IUPAC Name: 1-(4-bromothiophen-2-yl)ethan-1-one
Synonyms: 1-(4-bromothiophen-2-yl)ethanone
Molecular Weight (g/mol): 205.07
Molecular Formula: C6H5BrOS
InChi Key: HXVLWNKFMNRJED-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Br)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results