accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72126-54-6

IUPAC Name: 5-chloro-6-methylpyrazine-2,3-dicarbonitrile
Synonyms: 5-chloro-6-methylpyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 178.58
Molecular Formula: C7H3ClN4
InChi Key: FMSRCHYXAOIQFU-UHFFFAOYSA-N
SMILES: CC1=NC(C#N)=C(N=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results