accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 721457-32-5

IUPAC Name: (12-azidododecyl)phosphonic acid
Synonyms: 12-azidododecylphosphonic acid
Molecular Weight (g/mol): 291.33
Molecular Formula: C12H26N3O3P
InChi Key: WHRLXOVKIALVOF-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCCCCCCCCCCN=[N+]=[N-]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results