CAS RN 7228-38-8
IUPAC Name:
5-chloro-2H-1,3-benzodioxole
Synonyms:
5-chloro-2H-1,3-benzodioxole
Molecular Weight (g/mol):
156.57
Molecular Formula:
C7H5ClO2
InChi Key:
ODQPZHOXLYATLC-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2OCOC2=C1
1
–
1
of
1
results
1
Promotions Available
5-Chloro-1,3-benzodioxole, 98%
5-Chloro-1,3-benzodioxole, 98%
CAS: 7228-38-8 Molecular Formula: C7H5ClO2 Molecular Weight (g/mol): 156.565 MDL Number: MFCD00010842 InChI Key: ODQPZHOXLYATLC-UHFFFAOYSA-N PubChem CID: 138966 IUPAC Name: 5-chloro-1,3-benzodioxole SMILES: C1OC2=C(O1)C=C(C=C2)Cl