accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNH2

CAS RN 72299-67-3

IUPAC Name: 1,2,3,4-tetrahydroisoquinolin-6-amine
Synonyms: 1,2,3,4-tetrahydroisoquinolin-6-amine
Molecular Weight (g/mol): 148.21
Molecular Formula: C9H12N2
InChi Key: SAAFIVJVSQVSSW-UHFFFAOYSA-N
SMILES: NC1=CC2=C(CNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results